Learn More
Upchurch Scientific™ Back Pressure Regulators and Accessories
Placing back pressure regulator (usually 500 - 1,000 psi) between pump and injector can enhance pump's performance
Manufacturer:  Upchurch Scientific™ U607
 View more versions of this product
Catalog No. 05-704-222
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Back Pressure Regulator Assembly
Stainless-steel
129μL
100psi