Learn More
Idex Back Pressure Regulatos (BPR)
Enhance system performance through outgassing prevention
Manufacturer:  Idex U606
 View more versions of this product
Catalog No. 05-704-221
Add to cart
Description
Description
  • 1/4-28 flat-bottom ports
Select regulators with preset back pressures ranging from 5 to 1000 psi (0.3 to 69 bar). Each assembly comes with a replaceable, factory preset cartridge (except 5 and 20 psi versions).
Specifications
Specifications
Stainless Steel
Stainless Steel holder with ETFE flangeless ferrules and Delrin flangeless nuts for tubing
BPR Assembly
129 µL