Learn More
Idex Back Pressure Regulatos (BPR)
Enhance system performance through outgassing prevention
$171.35
Specifications
Type BPR Assembly
Includes Stainless Steel holder with ETFE flangeless ferrules and Delrin flangeless nuts for tubing
Material Stainless Steel
View More Specs
Products 1
Catalog Number Mfr. No. Swept Volume Price Quantity & Availability  
Catalog Number Mfr. No. Swept Volume Price Quantity & Availability  
05-704-223 Non-distribution product as customer accommodation.
View Documents
Idex
U608
99 µL
Each for $171.35
Add to cart
Description
Description
  • 1/4-28 flat-bottom ports
Select regulators with preset back pressures ranging from 5 to 1000 psi (0.3 to 69 bar). Each assembly comes with a replaceable, factory preset cartridge (except 5 and 20 psi versions).
Specifications
Specifications
BPR Assembly
Stainless Steel holder with ETFE flangeless ferrules and Delrin flangeless nuts for tubing
Stainless Steel