Learn More
Idex Barbed-to-Threaded Adapters
Easily connect soft-wall tubing to 10-32 coned, 1/4-28 flat-bottom, or 5/16-24 flat-bottom port
Manufacturer:  Idex P648
 View more versions of this product
Catalog No. 05-700-316
Add to cart
Description
Description
Slip soft-wall tubing over the barbed end then thread the adapter into a receiving port for a reliable low-pressure connection.
Specifications
Specifications
ETFE
Barbed-to-Threaded Adapter
4.76 mm
Connecting Soft Tubing to Standard 1/4-28 Internally-Threaded Ports