Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Idex Bottle Caps

Click to view available options
Neck Finish:
GL 38
GL 45
Color:
Aqua Blue
Red
For Use With (Equipment):
0.06 in. O.D. tubing
0.125 in. O.D. tubing

Bottle caps used for GL-38 and GL-45 tubing

Color Aqua Blue
No. per Case 1 Cs.
Type Cap
For Use With (Equipment) 0.125 in. O.D. tubing
Neck Finish GL 38