Learn More
Idex Cartridge Guard Column Kit
Guard columns and hardware to meet your system requirements
Manufacturer:  Idex C751
Catalog No. 05-702-55
Add to cart
Description
Description
  • Extend the life of analytical columns
  • Save on column replacement costs
  • Include 10-32 coned threads for easy connections
  • Flow path 100% biocompatible
  • Maximum psi 4000 (276 bar)