Upchurch Scientific™ Fingertight Ferrules

Manufacturer: Upchurch Scientific™ F148

 View more versions of this product

Catalog No. 05-700-54

  Check Availability
Add to Cart

    Specifications

    Material Kel-F
    Color Natural
    For Use With (Equipment) 190μm O.D. Tubing
    Product Type Fingertight ferrule

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit