Learn More
Idex Peristaltic Tubing Adapters
Connect 1/16 in. OD rigid/semi-rigid tubing to soft-wall tubing
$56.99
Specifications
Type Peristaltic Tubing Adapter
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
05-700-193 View Documents Idex
P757
Peristaltic to 1/16 in. O.D. Rigid Each for $56.99
Add to cart
Description
Description
Peristaltic tubing adapters feature specially designed nose to allow soft tubing to fit over the nose then be held in place by the retainer sleeve. To connect peristaltic tubing to rigid tubing with a different OD, order a flangeless fitting with 1/4-28 flat-bottom configuration.
Specifications
Specifications
Peristaltic Tubing Adapter