Learn More
Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16 in. OD Tubing, 10-32 Coned
Withstands up to 6000 psi.
Manufacturer:  Idex F300X
Catalog No. 05-700-11
Add to cart
Description
Description
Get the convenience of nut and ferrule built into one fitting. By using one-piece fittings there is no need to worry about the ferrule getting stuck in a receiving port.
Specifications
Specifications
Ferrule
For 0.06 in. O.D. Tubing
Undesignated
Two Piece Fingertight Double Winged Head Nut; 10 to 32 Coned
PEEK