Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Idex Flangeless Nuts: Compatible with 1/4-28, PEEK

No need to flange tubing

Manufacturer:  Idex P335X

 View more versions of this product

Catalog No. 05-700-396


Add to cart

Description

Description

Flangeless fitting eliminates the need to flange tubing. Just slide one of our Flangeless nuts onto your tubing with one of our Flangeless ferrules (coned end toward the nut) and finger tighten.

Specifications

Specifications

Natural
0.35 in.
0.89 cm
PEEK
Flangeless nut
9.4 cm
For 0.13 in. O.D. Tubing
0.61 x 0.37 in. (1.54 x 9.4 cm)
0.61 in.
1.54 cm
0.25 to 28 flat bottom
0.37 in.
10/Pk.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit