Learn More
Idex Flat-Bottom Fittings: ETFE Ferrules
No need to flange tubing
Manufacturer:  Idex P300N
 View more versions of this product
Catalog No. 05-700-101
Add to cart
Description
Description
Flangeless fitting eliminates the need to flange tubing. Just slide one of our Flangeless nuts onto your tubing with one of our Flangeless ferrules (coned end toward the nut) and finger tighten.
Specifications
Specifications
Standard Flat-Bottom Configurations
0.19 in.
Tefzel (ETFE)
Natural
0.19 in.
Not Applicable
Flangeless Ferrule
1/8 in.