Learn More
Idex Frits: PEEK
Extend filtration life and consistency with replaceable frits
Manufacturer:  Idex A701
 View more versions of this product
Catalog No. 05-704-184
Add to cart
Description
Description
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
Specifications
Specifications
PEEK
0.062 in.
0.16 cm
0.5 µm