Learn More
Idex High-Pressure Crosses
PEEK High-Pressure Unions, Tees, and Crosses hold up to 5000 psi and include FINGERTIGHT III fittings. Fittings work with any 1/16 in. OD tubing.
Manufacturer:  Idex P726
 View more versions of this product
Catalog No. 05-700-184
Add to cart
Specifications
Specifications
1/16 in. O.D. Tubing
Cross
1.25 mm
PEEK
0.05 in.
5000 psi