Learn More
Idex High-Pressure Crosses
PEEK High-Pressure Unions, Tees, and Crosses hold up to 5000 psi and include FINGERTIGHT III fittings. Fittings work with any 1/16 in. OD tubing.
$61.19 - $90.59
Specifications
Type Cross
For Use With (Equipment) 1/16 in. O.D. Tubing
Material PEEK
View More Specs
Products 2
Catalog Number Mfr. No. Pressure Rating Price Quantity & Availability  
Catalog Number Mfr. No. Pressure Rating Price Quantity & Availability  
05-700-184
View Documents
Idex
P726
5000 psi Each for $90.59
Add to cart
05-700-187
View Documents
Idex
P729
3500 psi Each for $61.19
Add to cart
Specifications
Specifications
Cross
1/16 in. O.D. Tubing
PEEK