Learn More
Idex High-Pressure Solvent Inline Filters
In-line Solvent Filters for running cleaner systems
Manufacturer:  Idex A431
Catalog No. 05-707-86
Add to cart
Description
Description
  • Help prevent particulates from clogging equipment
  • Parts made of stainless steel and PEEK
  • Replacement frits available
  • Max pressure 6000 psi (414 bar)
Specifications
Specifications
0.5μm
PEEK™