Learn More
Upchurch Scientific™ High-Pressure Unions
PEEK High-Pressure Unions, Tees, and Crosses hold up to 5000 psi and include FINGERTIGHT III fittings. Fittings work with any 1/16 in. OD tubing.
Manufacturer:  Upchurch Scientific™ U438
 View more versions of this product
Catalog No. 05-700-249
Add to cart
Specifications
Specifications
1/16 in. O.D. Tubing
Union
Near 0 µL
Stainless Steel
0.67 in.