Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Upchurch Scientific™ Inlet Filter

Filter solvents prevent damage to pumps and other system components

Manufacturer:  Upchurch Scientific™ A310

Catalog No. 05-707-26


Add to cart

Description

Description

  • Waters™ compatible
  • Stem included
  • Stainless steel
  • 10µm pore size
  • 0.32cm (0.125 in.) filter diameter
  • Analytical instrumentation
  • In vitro diagnostics
  • Biotechnology
  • Semiconductor/electronics
  • Laboratory

Specifications

Specifications

10μm
Stainless Steel
40mL/min.
Waters™ analytical HPLC systems
Each
Inlet Solvent Filter
To filter out particulate matter from the solvent that may otherwise damage hardware, for general use, preparative, specialized applications
Stem for 0.12 in. I.D. tubing
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit