Learn More
Upchurch Scientific™ Inlet Solvent Filters: End Fittings
Manufactured from Polyether ether ketone (PEEK). Upchurch Scientific Inlet Solvent Filters: End Fittings are used for Analytical HPLC Systems.
Manufacturer:  Upchurch Scientific™ A428X
 View more versions of this product
Catalog No. 05-707-83
  Check Availability
Add to cart
Specifications
Description
  • For Analytical HPLC System
  • 0.5µm PEEK filter end fittings
  • Black PEEK body
  • Swept volume of 5.7µL
Description & Specifications
Specifications
Replacement Filter End Fitting
PEEK
0.5μm
10 to 32 Coned