Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Upchurch Scientific™ Inlet Solvent Filters: End Fittings

Manufactured from Polyether ether ketone (PEEK). Upchurch Scientific Inlet Solvent Filters: End Fittings are used for Analytical HPLC Systems.

Manufacturer:  Upchurch Scientific™ A 429X

 View more versions of this product

Catalog No. 05-707-84


  Check Availability
Add to cart

Specifications

Description

  • For Analytical HPLC System
  • 0.2µm PEEK filter end fittings
  • Natural body
  • Swept volume of 6.9µL
  • Thread port: 10-32 Coned
Description & Specifications

Specifications

Replacement Filter End Fitting
PEEK
0.2μm
10 to 32 Coned
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit