Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Upchurch Scientific™ Inlet Solvent Filters: Filter Cups

Replacement parts for inlet solvent filters.

Manufacturer:  Upchurch Scientific™ A522X

 View more versions of this product

Catalog No. 05-707-56


  Check Availability
Add to cart

Specifications

Description

  • For use with A-551/A-553 filters
  • Material: Stainless Steel
  • Pore Size: 2µm
  • Flow Rate: 5mL/min.
Description & Specifications

Specifications

Replacement Filter Cup
Filters A-551/A-553
2μm
5mL/min.
Stainless Steel
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit