Learn More
Idex LiteTouch™ Nut, PEEK, 1/16 in. OD Tubing, 10-32 Coned
Secure a leak-tight connection
Manufacturer:  Idex LT110X
Catalog No. 05-700-274
Add to cart
Specifications
Specifications
Fits 1/16 in. O.D. tubing
10 to 32 UNF
LiteTouch Ferrule System
PEEK
Nut