Learn More
Idex Low-Pressure Flangeless Fittings
No need to flange tubing
Manufacturer:  Idex P200X
Catalog No. 05-700-76
Add to cart
Description
Description
Excellent for connecting rigid or semi-rigid tubing
Specifications
Specifications
ETFE
Blue
1/16 in. O.D. Tubing
Flangeless Ferrule