Learn More
Idex Low-Pressure Inline Filters Non-distribution product as customer accommodation.
These filters offer excellent protection and allow for tailoring to a variety of low-pressure applications.
Manufacturer:  Idex A415
 View more versions of this product
Catalog No. 05-707-42
Add to cart
Description
Description
Each filter is rated to 1,000 psi (68.9 bar). They work best when used with PTFE or ETFE tubing, but can be used with any appropriate size semi-rigid or rigid tubing.
  • Zero-dead volume connections
  • No wrenches required
  • For 1/16 in. or 1/8 in. OD tubing
When choosing alternate or replacement frits, select only those with an overall diameter of .200 in., including polymer ring.
Specifications
Specifications
2 μm
Stainless Steel
1/16 in. O.D. tubing