Learn More
Idex Low-Pressure Unions: Kel-F
Make solid connections in low-pressure applications
Manufacturer:  Idex P645
Catalog No. 05-700-170
Add to cart
Description
Description
  • Manufactured from inert polymers
  • Provide your own fittings to suit your application
  • 1/4-28 flat-bottom male thread union
  • 0.062 in. thru-hole diameter
  • PCTFE body
Specifications
Specifications
Kel-F
Natural
Union
80°C