Learn More
Idex Peek Low-Pressure Union Assembly
Make solid connections in low-pressure applications
Manufacturer:  Idex P720
 View more versions of this product
Catalog No. 05-700-370
Add to cart
Description
Description
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
  • Max pressure of 1000 psi (69 bar)
  • 0.030 in. thru-hole diameter
  • 0.41 µL swept volume
Specifications
Specifications
Natural
With 2 F-125/1 P-553, 6-32
PEEK
Union