Learn More
Idex Threaded Luer PEEK Adapter
Convenience of quick connect and disconnect of a luer connection
Manufacturer:  Idex P656
 View more versions of this product
Catalog No. 05-700-332
Add to cart
Description
Description
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Specifications
PEEK
To Female 10-32
0.05 in.
Natural
Luer Adapter
125°C