Learn More
Idex Threaded Luer Polypropylene Adapter
Convenience of quick connect and disconnect of a luer connection
Manufacturer:  Idex P618L
 View more versions of this product
Catalog No. 05-700-313
Add to cart
Description
Description
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Specifications
Natural
Female to Male
Luer Adapter
40°C
0.125 mm
Polypropylene
0.125 mm