Learn More
Idex Micro Tight™ Nuts
Fittings for very high pressure applications
Manufacturer:  Idex P416
Catalog No. 05-700-402
Add to cart
Description
Description
  • Ideal for UHPLC applications Quick component replacement with minimal down time
  • Max pressure of 15,000 psi (1035 bar)
  • Requires 4.0 in-lb (0.45 N-m) of torque
  • Natural PEEK
Specifications
Specifications
PEEK