Learn More
Idex Microbore Guard Columns
Extend filtration life and consistency with replaceable frits
Manufacturer:  Idex C128
Catalog No. 05-702-32
Add to cart
Description
Description
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
  • Frit volume 0.1 µL