Learn More
Idex Microtight Adapter
Easily connect capillary tubing with tubing sleeves
Manufacturer:  Idex P770
Catalog No. 05-700-371
Add to cart
Description
Description
  • Securely join together fluid pathway components
  • Holds tight connection even through vibration
  • Max pressure 4000 psi (276 bar)
Specifications
Specifications
1.58 to 0.63 mm