Learn More
Idex National Pipe Thread (NPT) Adapters
Maintain secure connections to 1/8 in. and 1/4 in. NPT ports even under high pressure
Manufacturer:  Idex U500
 View more versions of this product
Catalog No. 05-703-30
Add to cart
Description
Description
  • Securely join together fluid pathway components
  • Manufactured from PEEK polymer
  • Includes fittings
Specifications
Specifications
P-300/308 for 1/8 in. OD tubing
NPT Adapter
0.062 in.
PEEK
Red
0.25 in. Male NPT to 0.25-28 Female