Learn More
Idex PEEK Fitting Kit
PEEK Low-Pressure Unions, Tees, and Crosses hold up to 1000 psi (1/16 in. tubing), or to 500 psi (1/8 in. tubing). PEEK flangeless fittings included. Models 02008-00, -10, and -20 take 1/16 in. OD tubing; models 02008-04, -14, and -24 take 1/8 in. OD tubing.
Manufacturer:  Idex 1457
 View more versions of this product
Catalog No. 05-700-35
Add to cart
Description
Description
PEEK Fitting Kit contains convenient 1⁄4-28 threaded fittings for both 1⁄16 in. and 1⁄8 in. tubing:
  • 1⁄16 in. tubing: PEEK flangeless nuts (6), Tefzel™ flangeless ferrules (10), and PEEK 0.020 in. bore
    Tee (1), cross (1), and union (2). Also includes 10-32 to M6 adapters (2) and 10-32 to 1⁄4-28
    female adapters (2).
  • 1⁄8 in. tubing: PEEK flangeless nuts (6), Tefzel flangeless ferrules (10), and PEEK 0.050 in. bore
    Tee (1), cross (1), and union (2). Also includes 10-32 to M6 adapters (2) and 10-32 to 1⁄4-28
    female adapters (2).
Specifications
Specifications
PEEK
(20) ferrules, (6) Fingertight III nuts with ferrules, (2) metric adapters, (2) 10-32 female to 1/4-28 female adapters, (2) unions with 0.020-in- bore
High Pressure Fittings Kit