Learn More
Upchurch Scientific™ PEEK™ Micro Fingertight Ferrules
Manufacturer:  Upchurch Scientific™ F172
 View more versions of this product
Catalog No. 05-700-399
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Black
0.025 in. O.D. Tubing connect
Micro fingertight ferrule
PEEK