Learn More
Idex PEEK Microfilter Assembly
Combine multiple fluid streams into one under high operating pressures
Manufacturer:  Idex P775
Catalog No. 05-700-406
Add to cart
Description
Description
  • Securely join fluid pathway components Holds tight connection even through vibration
  • Manufactured from inert polymers
These multiport connectors are designed to maintain a secure joining of tubing or other fluid pathway components in mid- to high-pressure applications.
Specifications
Specifications
0.006 in.