Upchurch Scientific™ PEEK™ Sample Loops
Offer consistent volumes due to tight tolerances of extrusion processes
Manufacturer: Upchurch Scientific™ 1805
 View more versions of this product
Catalog No. 05-701-416
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Sample Loop
    Diameter (English) Inner 0.02 in.
    Diameter (Metric) Inner 0.05cm
    For Use With (Application) Analytical instrumentation, in-vitro diagnostics, biotechnology, laboratory
    Material PEEK Polymer
    Volume (Metric) Sample 50μL
  • Description & Specifications
Related Products
Product Content Feedback