Learn More
Idex PEEK Chromatography Tubing
An excellent replacement for stainless steel tubing
Manufacturer:  Idex 1531BL
 View more versions of this product
Catalog No. 05-701-12
Add to cart
Description
Description
Can be used in place of stainless steel tubing in most liquid analytical systems
  • Biocompatible, inert and easy to cut
  • Great for high-pressure applications
  • Wide choice of outer and inner diameters
  • Many sizes color-coded for easier inner diameter identification
Specifications
Specifications
50 ft.
Blue
7000 psi
PEEK Tubing
0.01 in.
For High-Pressure Applications