Learn More
Idex PEEK Chromatography Tubing
An excellent replacement for stainless steel tubing
Manufacturer:  Idex 1538XL
 View more versions of this product
Catalog No. 05-701-22
Add to cart
Description
Description
Can be used in place of stainless steel tubing in most liquid analytical systems
  • Biocompatible, inert and easy to cut
  • Great for high-pressure applications
  • Wide choice of outer and inner diameters
  • Many sizes color-coded for easier inner diameter identification
Specifications
Specifications
100 ft.
Natural
5000 psi
PEEK Tubing
0.04 in.
For High-Pressure Applications