Learn More
Upchurch Scientific™ PEEK Tubing
Manufacturer:  Upchurch Scientific™ 1542
 View more versions of this product
Catalog No. 05-701-26
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Can be used in place of stainless steel tubing in most liquid analytical systems
  • Biocompatible, inert and easy to cut
  • Great for high-pressure applications
  • Wide choice of outer and inner diameters
  • Many sizes color-coded for easier inner diameter identification
Specifications
Specifications
5 ft.
Natural
2000 psi
PEEK Tubing
0.01 in.
For High-Pressure Applications