Learn More
Idex PEEK™ Tubing Kit
An excellent replacement for stainless steel tubing
Manufacturer:  Idex 1326
Catalog No. 05-701-39
Add to cart
Description
Description
  • Flexible PEEK tubing is biocompatible and chemically inert
  • Easily cut to desired lengths
  • Smooth internal surface decreases turbulence
  • Color-coded for easier identification
  • Tubing cutter and six 5-ft long tubing sections
Specifications
Specifications
PEEK