Learn More
Idex Precolumn Filter
Precolumn filters offer secure protection for analytical columns
Manufacturer:  Idex A355
Catalog No. 05-701-399
Add to cart
Description
Description
  • Economical protection for larger columns and injections
  • Feature 10-32 coned ports for connection to 1/16 in. OD tubing
  • Replaceable stainless steel (SS) and PEEK frits
  • Max pressure 5000 psi (345 bar)
Specifications
Specifications
2μm
PEEK™