Learn More
Idex Precolumn Microfilters
Pressure rated to 4,000 psi
$188.37
Specifications
Type Microfilter
View More Specs
Products 3
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
05-707-105 View Documents Idex
M510
Capillary Tubing PEEK Frits (5 A-735) Each for $188.37
Add to cart
05-707-121 View Documents Idex
M500
Capillary Tubing Stainless Steel Frits (5 C-425) Each for $188.37
Add to cart
05-707-110 View Documents Idex
M560
1/16 in. Tubing PEEK Frits (5 A-735) Each for $188.37
Add to cart
Description
Description
Standard 10-32 male thread allows direct connection to your microbore or analytical column. The PEEK™ tubing used in the assembly of these units has a .005 in. I.D., virtually eliminating any mixing.
  • Direct connects to standard 10-32 ports
  • Only 0.5µL total theoretical void volume
  • Pressure rated to 4,000 psi (276 bar)
  • All included frits are 0.5µm porosity
  • Available with PEEK frits or stainless steel frits
  • Versions for use with 1/16 in. OD and 70-520µm O.D. capillary tubing
Specifications
Specifications
Microfilter