Upchurch Scientific™ Quick Connect Adapter

Manufacturer: Upchurch Scientific™ P677

 View more versions of this product

Catalog No. 05-700-366

  Check Availability
Add to Cart

    Specifications

    Material ETFE
    Type Luer Adapter
    For Use With Pair to female M6x1
    Color Black
    Temperature (Metric) Max 80°C
    Diameter (English) Inner 0.05 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit