Learn More
Idex Quick Connect Adapter
Convenience of quick connect and disconnect of a luer connection
Manufacturer:  Idex P679
 View more versions of this product
Catalog No. 05-700-368
Add to cart
Description
Description
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
Specifications
Specifications
Natural
ETFE
0.05 in.
Pair to Female 10-32
Female Luer Adapter
125°C