Learn More
Idex Semi-Prep Guard Cartridge
Guard columns and hardware to meet your system requirements
Manufacturer:  Idex C1035
Catalog No. 05-702-119
Add to cart
Description
Description
  • Extend the life of analytical columns
  • Save on column replacement costs
  • Include 10-32 coned threads for easy connections
  • Improves symmetry and plate count
  • Use 10-32 coned fittings to connect to 1/16 in. OD tubing