Learn More
Idex Soft Tubing Connectors
Connect 1/16 in. OD rigid/semi-rigid tubing to soft-wall tubing
Manufacturer:  Idex P767
Catalog No. 05-701-176
Add to cart
Description
Description
  • Retainer sleeve holds tubing tightly in place
  • For soft-wall tubing with up to 0.25 in. OD x 0.10 to 0.15 in. ID
  • 0.70 in. bore
  • Natural PEEK/Blue ETFE construction