Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Idex Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID

Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses

Manufacturer:  Idex U155

 View more versions of this product

Catalog No. 05-701-146


Add to cart

Description

Description

Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.

  • Precut 316 stainless steel tubing:
  • Ends are machine cut and polished
  • Edges are deburred inside and outside
  • Tubing is passivated
  • Inside is flushed with reagent-grade isopropanol
  • Color-coded banding for easy identification of the inner diameter
  • All stainless-steel tubing longer than 1m is coiled

Specifications

Specifications

316 Stainless Steel
30 cm
Red
1/16 in.
21,600 psi (1489 bar)
16 in.
Tubing
0.005 in.
0.125 mm
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit