Learn More
Idex Threaded Plugs and Caps
Plugs to close off unused ports in valves and multi-port connectors
  • Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Manufacturer:  Idex P554
 View more versions of this product
Catalog No. 05-700-363
Add to cart
Specifications
Specifications
10-32 Coned
Delrin (Acetal)
Plug
White