Learn More
Idex Threaded Plugs and Caps
Plugs to close off unused ports in valves and multi-port connectors
  • Seal 6-32, 10-32, 1/4-28, M6x1 and 5/16-24 threaded port
Manufacturer:  Idex P316
 View more versions of this product
Catalog No. 05-701-350
Add to cart
Specifications
Specifications
PFA
1/4-28 Flat Bottom
Plug
Natural