Learn More
Idex Threaded Port Adapters
Threaded port adapters help bring products with different thread configurations together
Manufacturer:  Idex P652
 View more versions of this product
Catalog No. 05-703-29
Add to cart
Specifications
Specifications
10-32 to 1/4-28
0.03 in.
1000 psi
PEEK
6.7 µL