Learn More
Idex Universal Column Couplers Non-distribution product as customer accommodation.
Great for reducing overall flow path volume
Manufacturer:  Idex U281
 View more versions of this product
Catalog No. 05-702-77
Add to cart
Description
Description
  • Couple two devices together
  • Lessen volume of flow path
  • 10-32 coned port connections
Column Couplers allow zero dead-volume connections between column and column, column and detector, or column and guard column.
Specifications
Specifications
10-32 Std. Ports, Kel-F w/ 0 0.020 in. Through-Hole
Universal Column Coupler
6.0 µL
Connect Column w/Internal 10-32 Threads to Precolumn or Guard Column w/10-32 Threads